Search
Search

sog| Gene Symbol: sog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272647.2 | NP_001259576.1 | 4 - 5 | 1669 | 66 | |
| NM_001272649.2 | NP_001259578.1 | 3 - 4 | 1351 | 66 | ||
| NM_057388.4 | NP_476736.1 | 3 - 4 | 1351 | 66 | ||
| GenBank mRNA | BT053679.1 | - | 4 - 5 | 1669 | 66 | |
| U18774.1 | - | 3 - 4 | 1696 | 66 | ||
Set Membership: |
|