Search
Search

Nup154| Gene Symbol: Nup154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298898.1 | NP_001285827.1 | 11 - 12 | 4126 | 74 | |
| NM_057939.4 | NP_477287.1 | 11 - 12 | 4126 | 74 | ||
| NM_057940.4 | NP_477288.3 | 11 - 12 | 4126 | 74 | ||
| GenBank mRNA | AF160944.1 | - | 11 - 12 | 4092 | 74 | |
Set Membership: |
|