Search
Search

DIP-theta| Gene Symbol: DIP-theta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164653.3 | NP_723103.1 | 10 - 11 | 1685 | 67 | |
| GenBank mRNA | BT150446.1 | - | 10 - 11 | 1678 | 67 | |
Set Membership: |
|