Search
Search

kat-60L1| Gene Symbol: kat-60L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170050.1 | NP_001163521.1 | 3 - 4 | 1317 | 60 | |
| NM_001170051.1 | NP_001163522.1 | 3 - 4 | 1163 | 60 | ||
| NM_001170052.1 | NP_001163523.1 | 3 - 4 | 1329 | 60 | ||
| NM_001170053.1 | NP_001163524.1 | 3 - 4 | 1151 | 60 | ||
| NM_001170054.1 | NP_001163525.1 | 3 - 4 | 1329 | 60 | ||
| NM_001170055.1 | NP_001163526.1 | 3 - 4 | 1163 | 60 | ||
| NM_141329.3 | NP_649586.1 | 3 - 4 | 1151 | 60 | ||
| NM_169105.2 | NP_731004.1 | 3 - 4 | 1317 | 60 | ||
| GenBank mRNA | AY051591.1 | - | 3 - 4 | 1151 | 60 | |
| BT001274.1 | - | 3 - 4 | 1317 | 60 | ||
| BT023721.1 | - | 3 - 4 | 1252 | 60 | ||
Set Membership: |
|