Search
Search

Osi17| Gene Symbol: Osi17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275377.1 | NP_001262306.1 | 5 - 6 | 2628 | 79 | |
| NM_141381.3 | NP_649638.2 | 4 - 5 | 1915 | 79 | ||
| GenBank mRNA | AY118624.1 | - | 4 - 5 | 1912 | 79 | |
Set Membership: |
|