Search
Search

SP1029| Gene Symbol: SP1029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276136.1 | NP_001263065.1 | 2 - 3 | 759 | 59 | |
| NM_001300651.1 | NP_001287580.1 | 2 - 3 | 542 | 59 | ||
| NM_144361.2 | NP_652618.1 | 2 - 3 | 587 | 59 | ||
| NM_170405.2 | NP_733284.1 | 3 - 4 | 882 | 59 | ||
| NM_170406.2 | NP_733285.1 | 2 - 3 | 719 | 59 | ||
| GenBank mRNA | BT133378.1 | - | 2 - 3 | 581 | 59 | |
Set Membership: |
|