Search
Search

Dhc98D| Gene Symbol: Dhc98D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170288.1 | NP_001163759.1 | 14 - 15 | 5403 | 60 | |
| NM_001276107.1 | NP_001263036.1 | 8 - 9 | 3085 | 60 | ||
| NM_079817.3 | NP_524541.3 | 15 - 16 | 5424 | 60 | ||
Set Membership: |
|