Search
Search

chp| Gene Symbol: chp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276205.1 | NP_001263134.1 | 3 - 4 | 1399 | 67 | |
| NM_001276206.1 | NP_001263135.1 | 3 - 4 | 1420 | 67 | ||
| NM_001276207.1 | NP_001263136.1 | 3 - 4 | 1399 | 67 | ||
| NM_001276208.1 | NP_001263137.1 | 3 - 4 | 1420 | 67 | ||
| NM_079866.3 | NP_524605.1 | 3 - 4 | 1420 | 67 | ||
| GenBank mRNA | BT126010.1 | - | 3 - 4 | 1198 | 67 | |
Set Membership: |
|