Search
Search

ppk10| Gene Symbol: ppk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038805.2 | NP_001033894.1 | 10 - 11 | 1535 | 87 | |
| GenBank mRNA | AY226541.1 | - | 10 - 11 | 1340 | 87 | |
Set Membership: |
|