Search
Search

cryaba| Gene Symbol: cryaba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131157.1 | NP_571232.1 | 1 - 2 | 221 | 77 | |
| GenBank mRNA | AF159089.2 | - | 1 - 2 | 221 | 77 | |
| BC162184.1 | - | 1 - 2 | 220 | 77 | ||
| BC162206.1 | - | 1 - 2 | 220 | 77 | ||
Set Membership: |
|