Search
Search

nfyc| Gene Symbol: nfyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199639.1 | NP_955933.1 | 7 - 8 | 734 | 64 | |
| XM_009294374.3 | XP_009292649.1 | 7 - 8 | 779 | 64 | ||
| XM_009294375.3 | XP_009292650.1 | 7 - 8 | 1559 | 64 | ||
| XM_009294376.3 | XP_009292651.1 | 7 - 8 | 1556 | 64 | ||
| XM_009294377.3 | XP_009292652.1 | 7 - 8 | 776 | 64 | ||
| XM_009294378.3 | XP_009292653.1 | 7 - 8 | 753 | 64 | ||
| XM_009294379.3 | XP_009292654.1 | 7 - 8 | 1556 | 64 | ||
| XM_021467995.1 | XP_021323670.1 | 7 - 8 | 1553 | 64 | ||
| GenBank mRNA | BC045364.1 | - | 7 - 8 | 734 | 64 | |
| BC065645.1 | - | 7 - 8 | 665 | 64 | ||
| BC164181.1 | - | 6 - 7 | 664 | 64 | ||
Set Membership: |
|