Search
Search

aimp2| Gene Symbol: aimp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213338.1 | NP_998503.1 | 1 - 2 | 233 | 101 | |
| GenBank mRNA | BC053178.1 | - | 1 - 2 | 233 | 101 | |
| BC164848.1 | - | 1 - 2 | 163 | 101 | ||
Set Membership: |
|