Search
Search

crlf1a| Gene Symbol: crlf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002650.1 | NP_001002650.1 | 5 - 6 | 1036 | 86 | |
| XM_009298725.3 | XP_009297000.1 | 6 - 7 | 1370 | 86 | ||
| XM_017358225.2 | XP_017213714.1 | 6 - 7 | 1368 | 86 | ||
| GenBank mRNA | BC076526.1 | - | 5 - 6 | 1066 | 86 | |
| BC164710.1 | - | 5 - 6 | 825 | 86 | ||
Set Membership: |
|