Search
Search

iqch| Gene Symbol: iqch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045129.1 | NP_001038594.1 | 5 - 6 | 519 | 62 | |
| XM_005159132.4 | XP_005159189.1 | 5 - 6 | 581 | 62 | ||
| XM_005159133.4 | XP_005159190.1 | 5 - 6 | 581 | 62 | ||
| GenBank mRNA | BC161661.1 | - | 5 - 6 | 520 | 62 | |
Set Membership: |
|