Search
Search

cyp11c1| Gene Symbol: cyp11c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080204.1 | NP_001073673.1 | 2 - 3 | 510 | 81 | |
| XM_017351368.2 | XP_017206857.1 | 6 - 7 | 1749 | 81 | ||
| XM_017351369.2 | XP_017206858.1 | 7 - 8 | 1796 | 81 | ||
| XM_017351370.2 | XP_017206859.1 | 6 - 7 | 1613 | 81 | ||
| XM_021466759.1 | XP_021322434.1 | 7 - 8 | 1793 | 81 | ||
| GenBank mRNA | BC122212.1 | - | 1 - 2 | 268 | 81 | |
| BC155806.1 | - | 5 - 6 | 1028 | 81 | ||
| DQ650710.1 | - | 2 - 3 | 510 | 81 | ||
Set Membership: |
|