Search
Search

herpud2| Gene Symbol: herpud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200188.1 | NP_956482.1 | 2 - 3 | 421 | 128 | |
| XM_021466640.1 | XP_021322315.1 | 2 - 3 | 478 | 128 | ||
| GenBank mRNA | BC045865.1 | - | 2 - 3 | 421 | 128 | |
| BC066585.1 | - | 2 - 3 | 404 | 128 | ||
Set Membership: |
|