Search
Search

pdcd10a| Gene Symbol: pdcd10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200555.2 | NP_956849.2 | 5 - 6 | 566 | 76 | |
| XM_005158980.4 | XP_005159037.1 | 5 - 6 | 515 | 76 | ||
| GenBank mRNA | BC056552.1 | - | 5 - 6 | 540 | 76 | |
| EH548629.1 | - | 5 - 6 | 605 | 76 | ||
Set Membership: |
|