Search
Search

macroh2a2| Gene Symbol: macroh2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025502.1 | NP_001020673.1 | 4 - 5 | 645 | 102 | |
| XM_009307097.1 | XP_009305372.1 | 4 - 5 | 525 | 102 | ||
| GenBank mRNA | BC097115.1 | - | 4 - 5 | 645 | 102 | |
Set Membership: |
|