Search
Search

dcdc2b| Gene Symbol: dcdc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037689.2 | NP_001032778.2 | 8 - 9 | 856 | 80 | |
| XM_017358763.2 | XP_017214252.1 | 10 - 11 | 1222 | 80 | ||
| XM_017358764.2 | XP_017214253.1 | 10 - 11 | 1183 | 80 | ||
| GenBank mRNA | BC108062.1 | - | 8 - 9 | 867 | 80 | |
Set Membership: |
|