Search
Search

atic| Gene Symbol: atic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082796.2 | NP_001076265.2 | 4 - 5 | 418 | 63 | |
| GenBank mRNA | BC045385.1 | - | 4 - 5 | 466 | 63 | |
| BC163887.1 | - | 4 - 5 | 418 | 63 | ||
Set Membership: |
|