Search
Search

bco2b| Gene Symbol: bco2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040312.1 | NP_001035402.1 | 10 - 11 | 1493 | 95 | |
| GenBank mRNA | BC115259.1 | - | 10 - 11 | 1493 | 95 | |
Set Membership: |
|