Search
Search

wnt2| Gene Symbol: wnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130950.1 | NP_571025.1 | 3 - 4 | 751 | 79 | |
| GenBank mRNA | BC056277.1 | - | 3 - 4 | 766 | 79 | |
| U51266.1 | - | 3 - 4 | 751 | 79 | ||
Set Membership: |
|