Search
Search

mxe| Gene Symbol: mxe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182867.1 | NP_878287.1 | 7 - 8 | 1052 | 104 | |
| XM_005167664.4 | XP_005167721.2 | 8 - 9 | 1184 | 104 | ||
| GenBank mRNA | AJ544827.1 | - | 7 - 8 | 1052 | 104 | |
Set Membership: |
|