Search
Search

tardbpl| Gene Symbol: tardbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198364.2 | NP_938178.1 | 3 - 4 | 935 | 55 | |
| XM_021469762.1 | XP_021325437.1 | 3 - 4 | 1003 | 55 | ||
| GenBank mRNA | BC044405.1 | - | 3 - 4 | 804 | 55 | |
| BC164253.1 | - | 3 - 4 | 575 | 55 | ||
Set Membership: |
|