Search
Search

surf6| Gene Symbol: surf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201065.1 | NP_957359.1 | 2 - 3 | 262 | 71 | |
| XM_005171804.4 | XP_005171861.1 | 2 - 3 | 71 | |||
| GenBank mRNA | AY648750.1 | - | 2 - 3 | 71 | ||
| BC053193.1 | - | 2 - 3 | 262 | 71 | ||
| BC164187.1 | - | 1 - 2 | 129 | 71 | ||
Set Membership: |
|