Search
Search

adgrg1| Gene Symbol: adgrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020481.1 | NP_001018317.1 | 12 - 13 | 1825 | 70 | |
| XM_005166389.4 | XP_005166446.1 | 13 - 14 | 2172 | 70 | ||
| GenBank mRNA | AB193557.1 | - | 12 - 13 | 1825 | 70 | |
Set Membership: |
|