Search
Search

edf1| Gene Symbol: edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200745.1 | NP_957039.1 | 3 - 4 | 381 | 84 | |
| GenBank mRNA | BC059541.1 | - | 3 - 4 | 381 | 84 | |
| BC071480.1 | - | 3 - 4 | 396 | 84 | ||
| BC159201.1 | - | 3 - 4 | 369 | 84 | ||
Set Membership: |
|