Search
Search

rps13| Gene Symbol: rps13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002079.1 | NP_001002079.1 | 5 - 6 | 432 | 83 | |
| GenBank mRNA | BC072552.1 | - | 5 - 6 | 432 | 83 | |
| BC163978.1 | - | 5 - 6 | 456 | 83 | ||
Set Membership: |
|