Search
Search

smyd1b| Gene Symbol: smyd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039636.1 | NP_001034725.1 | 1 - 2 | 120 | 100 | |
| XM_021478292.1 | XP_021333967.1 | 1 - 2 | 226 | 100 | ||
| GenBank mRNA | BC115351.1 | - | 1 - 2 | 125 | 100 | |
| BC163095.1 | - | 1 - 2 | 209 | 100 | ||
| DQ323979.1 | - | 1 - 2 | 120 | 100 | ||
| DQ323980.1 | - | 1 - 2 | 120 | 100 | ||
Set Membership: |
|