Search
Search

tssk6| Gene Symbol: tssk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044845.1 | NP_001038310.1 | 1 | 744 | 64 | |
| GenBank mRNA | BC171387.1 | - | 1 | 822 | 64 | |
| BC171391.1 | - | 1 | 822 | 64 | ||
Set Membership: |
|