Search
Search

zfr| Gene Symbol: zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199558.2 | NP_955852.2 | 9 - 10 | 1740 | 69 | |
| GenBank mRNA | BC055641.1 | - | 995 | 69 | ||
| BC059198.1 | - | 9 - 10 | 1717 | 69 | ||
Set Membership: |
|