Search
Search

sec22a| Gene Symbol: sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199698.1 | NP_955992.1 | 5 - 6 | 785 | 84 | |
| XM_021478609.1 | XP_021334284.1 | 5 - 6 | 811 | 84 | ||
| GenBank mRNA | BC049530.1 | - | 5 - 6 | 785 | 84 | |
Set Membership: |
|