Search
Search

aldoca| Gene Symbol: aldoca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029952.1 | NP_001025123.1 | 5 - 6 | 622 | 73 | |
| XM_009291564.3 | XP_009289839.1 | 5 - 6 | 581 | 73 | ||
| GenBank mRNA | BC098624.1 | - | 5 - 6 | 622 | 73 | |
Set Membership: |
|