Search
Search

rad51ap1
si:ch211-214j24.1| Gene Symbol: rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044338.2 | NP_001037803.2 | 8 - 9 | 1018 | 131 | |
| XM_009299961.3 | XP_009298236.1 | 8 - 9 | 1029 | 131 | ||
| GenBank mRNA | BC133972.1 | - | 8 - 9 | 1018 | 131 | |
| Gene Symbol: si:ch211-214j24.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI673060.1 | - | 2 - 3 | 200 | 131 | |
Set Membership: |
|