Search
Search

st6galnac4| Gene Symbol: st6galnac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045370.1 | NP_001038835.1 | 5 - 6 | 751 | 83 | |
| GenBank mRNA | AJ868430.1 | - | 3 - 4 | 666 | 83 | |
| BC122199.1 | - | 5 - 6 | 751 | 83 | ||
Set Membership: |
|