Search
Search

saraf| Gene Symbol: saraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076589.1 | NP_001070057.1 | 5 - 6 | 1171 | 116 | |
| XM_005159818.4 | XP_005159875.1 | 5 - 6 | 1201 | 116 | ||
| GenBank mRNA | BC124302.1 | - | 5 - 6 | 1171 | 116 | |
Set Membership: |
|