Search
Search

dachd| Gene Symbol: dachd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080673.1 | NP_001074142.1 | 6 - 8 | 1470 | 67 | |
| XM_005167750.4 | XP_005167807.1 | 6 - 7 | 1465 | 67 | ||
| GenBank mRNA | BC129504.1 | - | 6 - 8 | 1470 | 67 | |
Set Membership: |
|