Search
Search

pi4kab| Gene Symbol: pi4kab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113342.1 | NP_001106813.1 | 13 - 14 | 1600 | 86 | |
| GenBank mRNA | BC155592.1 | - | 13 - 14 | 1600 | 86 | |
Set Membership: |
|