Search
Search

nos2b| Gene Symbol: nos2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113501.1 | NP_001106973.1 | 16 - 17 | 2067 | 71 | |
| XM_005157694.4 | XP_005157751.1 | 8 - 9 | 830 | 71 | ||
| XM_009291655.3 | XP_009289930.1 | 15 - 16 | 3124 | 71 | ||
| XM_017350981.2 | XP_017206470.1 | 15 - 16 | 3122 | 71 | ||
| GenBank mRNA | AM749802.1 | - | 16 - 17 | 2067 | 71 | |
Set Membership: |
|