Search
Search

slc35c2| Gene Symbol: slc35c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212643.1 | NP_997808.1 | 5 - 6 | 533 | 93 | |
| XM_009296719.3 | XP_009294994.1 | 5 - 6 | 682 | 93 | ||
| XM_009296720.3 | XP_009294995.1 | 5 - 6 | 705 | 93 | ||
| GenBank mRNA | BC045291.1 | - | 5 - 6 | 533 | 93 | |
| BC068979.1 | - | 5 - 6 | 607 | 93 | ||
Set Membership: |
|