Search
Search

LPIN1| Gene Symbol: LPIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502170.6 | XP_001502220.4 | 277 | 84 | ||
| XM_005600211.3 | XP_005600268.1 | 414 | 84 | |||
| XM_005600212.3 | XP_005600269.1 | 415 | 84 | |||
| XM_005600213.3 | XP_005600270.1 | 493 | 84 | |||
| XM_005600214.3 | XP_005600271.1 | 414 | 84 | |||
| XM_005600215.3 | XP_005600272.1 | 391 | 84 | |||
| XM_005600216.3 | XP_005600273.1 | 415 | 84 | |||
| XM_023619510.1 | XP_023475278.1 | 339 | 84 | |||
| XM_023619511.1 | XP_023475279.1 | 223 | 84 | |||
Set Membership: |
|