Search
Search

KAT6A| Gene Symbol: KAT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630432.1 | XP_023486200.1 | 2426 | 64 | ||
| XM_023630433.1 | XP_023486201.1 | 2405 | 64 | |||
| XM_023630434.1 | XP_023486202.1 | 2310 | 64 | |||
| XM_023630435.1 | XP_023486203.1 | 5203 | 64 | |||
Set Membership: |
|