Search
Search

HTRA2| Gene Symbol: HTRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500046.5 | XP_001500096.1 | 587 | 74 | ||
| XM_014731077.2 | XP_014586563.1 | 589 | 74 | |||
| XM_023618835.1 | XP_023474603.1 | 589 | 74 | |||
Set Membership: |
|