Search
Search

ZDHHC5| Gene Symbol: ZDHHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729521.2 | XP_014585007.2 | 1892 | 88 | ||
| XM_023654084.1 | XP_023509852.1 | 2071 | 88 | |||
| XM_023654085.1 | XP_023509853.1 | 1845 | 88 | |||
| XM_023654086.1 | XP_023509854.1 | 541 | 88 | |||
Set Membership: |
|