Search
Search

RSRC2| Gene Symbol: RSRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647241.1 | XP_023503009.1 | 865 | 76 | ||
| XM_023647243.1 | XP_023503011.1 | 884 | 76 | |||
| XM_023647244.1 | XP_023503012.1 | 992 | 76 | |||
| XM_023647245.1 | XP_023503013.1 | 763 | 76 | |||
| GenBank mRNA | HQ890085.1 | - | 2 - 3 | 323 | 76 | |
Set Membership: |
|