Search
Search

GDPD5| Gene Symbol: GDPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645700.1 | XP_023501468.1 | 2369 | 97 | ||
| XM_023645701.1 | XP_023501469.1 | 1717 | 97 | |||
| XM_023645702.1 | XP_023501470.1 | 1623 | 97 | |||
| XM_023645703.1 | XP_023501471.1 | 2279 | 97 | |||
| XM_023645704.1 | XP_023501472.1 | 2368 | 97 | |||
| XM_023645705.1 | XP_023501473.1 | 2371 | 97 | |||
| XM_023645706.1 | XP_023501474.1 | 2370 | 97 | |||
| XM_023645707.1 | XP_023501475.1 | 2241 | 97 | |||
| XM_023645708.1 | XP_023501476.1 | 2240 | 97 | |||
Set Membership: |
|