Search
Search

TMCO3| Gene Symbol: TMCO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621890.1 | XP_023477658.1 | 1160 | 64 | ||
| XM_023621891.1 | XP_023477659.1 | 1064 | 64 | |||
| XM_023621892.1 | XP_023477660.1 | 1160 | 64 | |||
| XM_023621893.1 | XP_023477661.1 | 1160 | 64 | |||
| XM_023621894.1 | XP_023477662.1 | 1160 | 64 | |||
Set Membership: |
|