Search
Search
| Gene Symbol: IWS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023622655.1 | XP_023478423.1 | 371 | 81 | ||
| XM_023622656.1 | XP_023478424.1 | 386 | 81 | |||
| XM_023622657.1 | XP_023478425.1 | 371 | 81 | |||
| XM_023622658.1 | XP_023478426.1 | 371 | 81 | |||
| XM_023622659.1 | XP_023478427.1 | 371 | 81 | |||
| XM_023622660.1 | XP_023478428.1 | 371 | 81 | |||
| XM_023622661.1 | XP_023478429.1 | 386 | 81 | |||
Set Membership: |
|