Search
Search

UNC50| Gene Symbol: UNC50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618606.1 | XP_023474374.1 | 423 | 74 | ||
| XM_023618607.1 | XP_023474375.1 | 426 | 74 | |||
| XM_023618608.1 | XP_023474376.1 | 426 | 74 | |||
| GenBank mRNA | JL618539.1 | - | 3 - 4 | 366 | 74 | |
Set Membership: |
|