Search
Search
| Gene Symbol: SDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619466.1 | XP_023475234.1 | 218 | 100 | ||
| XM_023619467.1 | XP_023475235.1 | 305 | 100 | |||
| XM_023619468.1 | XP_023475236.1 | 421 | 100 | |||
Set Membership: |
|